missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC52 Antibody (1037552), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne FAB10260C
This item is not returnable.
View return policy
Description
LRRC52 Monoclonal antibody specifically detects LRRC52 in Human samples. It is validated for Flow Cytometry
Specifications
| LRRC52 | |
| Monoclonal | |
| PerCP | |
| FLJ25811, leucine rich repeat containing 52, leucine-rich repeat-containing protein 52 | |
| Human embryonic kidney cell HEK293-derived human LRRC52, Ser24-Asp244, Accession # Q8N7C0 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1037552 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 440699 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction