missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC6 Antibody (OTI7E9), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72542AF700
This item is not returnable.
View return policy
Description
LRRC6 Monoclonal specifically detects LRRC6 in Human samples. It is validated for Immunohistochemistry.
Specifications
| LRRC6 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LRRC6 | |
| Full length recombinant protein of human LRRC6 (NP_036604) produced in HEK293T cells. | |
| 0.1 mL | |
| 23639.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI7E9 | |
| Immunohistochemistry | |
| leucine rich repeat containing 6, Leucine-rich testis-specific protein, LRTPleucine-rich repeat-containing protein 6, Testis-specific leucine-rich repeat protein, TSLRPtestis specific leucine rich repeat protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction