missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRRC70 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15771001 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15771001 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15771001 Supplier Invitrogen™ Supplier No. PA553056

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PLSSLIHLQA NSNPWECNCK LLGLRDWLAS SAITLNIYCQ NPPSMRGRAL RYINITNCVT SSINVSRAWA VVKSPHIHHK TTALMMAWHK VTTNGSPLEN TETENITFWE RIPTSPAGRF FQENAFGNPL ETTAVLPVQI QL Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 19%.

Renders cells highly sensitive to the activation by cytokines and lipopolysaccharide (LPS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC70
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC70
Gene Accession No. Q7Z2Q7
Gene Alias leucine rich repeat containing 70; leucine-rich repeat-containing protein 70; LRRC70; SLRN; Synleurin; UNQ1891/PRO4337
Gene Symbols LRRC70
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC70. Recombinant protein control fragment (Product #RP-90563).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100130733
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.