missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC8A Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32158AF750
This item is not returnable.
View return policy
Description
LRRC8A Polyclonal specifically detects LRRC8A in Human, Mouse samples. It is validated for Western Blot.
Specifications
| LRRC8A | |
| Polyclonal | |
| Western Blot | |
| AGM5, FLJ10337, FLJ41617, KIAA1437leucine rich repeat containing 8, leucine rich repeat containing 8 family, member A, leucine-rich repeat-containing protein 8A, LRRC8 | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LRRC8A | |
| The immunogen this antibody was made to, maps to a region between residue 760 to 810 of human Leucine Rich Repeat Containing 8 Family, Member A using the numbering given in entry NP_001120716.1 (GeneID 56262). | |
| 100 μL | |
| 56262.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction