missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC8A Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32158C
This item is not returnable.
View return policy
Description
LRRC8A Polyclonal specifically detects LRRC8A in Human, Mouse samples. It is validated for Western Blot.
Specifications
| LRRC8A | |
| Polyclonal | |
| Western Blot | |
| AGM5, FLJ10337, FLJ41617, KIAA1437leucine rich repeat containing 8, leucine rich repeat containing 8 family, member A, leucine-rich repeat-containing protein 8A, LRRC8 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 56262 | |
| Store at 4C in the dark. |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LRRC8A | |
| The immunogen this antibody was made to, maps to a region between residue 760 to 810 of human Leucine Rich Repeat Containing 8 Family, Member A using the numbering given in entry NP_001120716.1 (GeneID 56262). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction