missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC8B Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97498AF350
This item is not returnable.
View return policy
Description
LRRC8B Polyclonal specifically detects LRRC8B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| LRRC8B | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| KIAA0231TA-LRRPleucine-rich repeat-containing protein 8B, leucine rich repeat containing 8 family, member B, MGC42220, T cell activation leucine repeat rich protein, TALRRP, T-cell activation leucine repeat-rich protein | |
| Produced in rabbits immunized with E. coli-derived Human LRRC8B fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 23507 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction