missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC8B Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97498APC
This item is not returnable.
View return policy
Description
LRRC8B Polyclonal antibody specifically detects LRRC8B in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| LRRC8B | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| KIAA0231TA-LRRPleucine-rich repeat-containing protein 8B, leucine rich repeat containing 8 family, member B, MGC42220, T cell activation leucine repeat rich protein, TALRRP, T-cell activation leucine repeat-rich protein | |
| Produced in rabbits immunized with E. coli-derived Human LRRC8B fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 23507 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction