missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC8B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99221-50ul
This item is not returnable.
View return policy
Description
LRRC8B Polyclonal antibody specifically detects LRRC8B in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| LRRC8B | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 uL/mg of lysate | |
| KIAA0231TA-LRRPleucine-rich repeat-containing protein 8B, leucine rich repeat containing 8 family, member B, MGC42220, T cell activation leucine repeat rich protein, TALRRP, T-cell activation leucine repeat-rich protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human LRRC8B. | |
| 50 μg | |
| Stem Cell Markers | |
| 23507 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction