Learn More
Invitrogen™ LRRC9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139957
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KHLLQILEKG FKDSETSKLP LKKEAIIVSN SLSISECPRI EFLQQKHKDE KKISLKHELF RHGILLITKV FLGQSVQAHE KESISQSNYP MVNSVFIPRK Y Highest antigen sequence indentity to the following orthologs: Mouse - 67%, Rat - 67%.
LRRC9 is a protein coding gene.
Specifications
| LRRC9 | |
| Polyclonal | |
| Unconjugated | |
| LRRC9 | |
| FLJ46156; leucine rich repeat containing 9; LRRC9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 341883 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZRR7 | |
| LRRC9 | |
| Recombinant protein corresponding to Human LRRC9. Recombinant protein control fragment (Product #RP-109644). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.