missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRIQ3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546528
This item is not returnable.
View return policy
Description
Peptide sequence: LRTFSDIDKY YTEQKKQEYH KEKVRVVAMA QVARERVRVA VNEHLNQKKY.
The function of this protein remains unknown.
Specifications
| LRRIQ3 | |
| Polyclonal | |
| Unconjugated | |
| LRRIQ3 | |
| leucine rich repeat containing 44; leucine rich repeats and IQ motif containing 3; leucine-rich repeat and IQ domain-containing protein 3; Leucine-rich repeat-containing protein 44; leucine-rich repeats and IQ motif containing 3; LRRC44; Lrriq3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 127255 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A6PVS8 | |
| LRRIQ3 | |
| Synthetic peptide directed towards the C-terminal of human LRRIQ3 (aa 417-466). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction