Learn More
Invitrogen™ LRRK2 Monoclonal Antibody (N231B/34), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545641
Description
81% identical in mouse, 80% identical in rat. <30% identity with LRRK1. 10 μg/mL of MA5-45641 was sufficient for detection of LRRK2/Dardarin in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects >200kDa. This antibody was formerly sold as clone S231B-34.
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. Mutations in LRRK2 have been associated with Parkinson's disease. LRRK2 is a member of the leucine-rich repeat kinase family and encodes a protein with an ankryin repeat region, a leucine-rich repeat domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, a MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane.
Specifications
| LRRK2 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4 | |
| Q5S007 | |
| LRRK2 | |
| Fusion protein amino acids 841-960 of human LRRK2. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N231B/34 | |
| APC | |
| LRRK2 | |
| 4921513O20Rik; 9330188B09Rik; augmented in rheumatoid arthritis 17; AURA17; AW561911; BC 300-268; cI-46; D630001M17Rik; DARDARIN; DKFZp434H2111; FLJ45829; Gm927; leucine rich repeat kinase 2; leucine-rich repeat kinase 2; leucine-rich repeat serine/threonine-protein kinase 2; LRR kinase 2; LRRK 2; Lrrk2; LRRK-2; PARK8; RIPK7; ROCO2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 120892, 300160 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.