missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRN1/NLRR-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 588.00
Specifications
| Antigen | LRRN1/NLRR-1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18429520
|
Novus Biologicals
NBP1-83896-25ul |
25 μL |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18003444
|
Novus Biologicals
NBP1-83896 |
0.1 mL |
€ 588.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRRN1/NLRR-1 Polyclonal specifically detects LRRN1/NLRR-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| LRRN1/NLRR-1 | |
| Polyclonal | |
| Rabbit | |
| Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 57633 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NSNKTNIRFMEPLSMFCAMPPEYKGHQVKEVLIQDSSEQCLPMISHDSFPNRLNVDIGTTVFLDCRAMAEPEPEIYWVTPIGNKITVETLSDKYKLSSEGTLEISNIQIEDSGRYTCVAQNVQGAD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| fibronectin type III, immunoglobulin and leucine rich repeat domains 3, FIGLER3, KIAA1497, leucine rich repeat neuronal 1, leucine-rich repeat neuronal protein 1, Neuronal leucine-rich repeat protein 1, NLRR-1 | |
| LRRN1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title