missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRTM1 Antibody (OTI5H9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72543CL1
This item is not returnable.
View return policy
Description
LRRTM1 Monoclonal antibody specifically detects LRRTM1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| LRRTM1 | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ32082, leucine rich repeat transmembrane neuronal 1, leucine-rich repeat transmembrane neuronal 1 protein, leucine-rich repeat transmembrane neuronal protein 1 | |
| Human recombinant protein fragment corresponding to amino acids 182-422 of human LRRTM1 (NP_849161) produced in E.coli. | |
| 0.1 mL | |
| Neuroscience | |
| 347730 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Immunohistochemistry, Western Blot | |
| OTI5H9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction