missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRTM2 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98780FR
This item is not returnable.
View return policy
Description
LRRTM2 Polyclonal specifically detects LRRTM2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LRRTM2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| leucine-rich repeat transmembrane neuronal protein 2, leucine rich repeat transmembrane neuronal 2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human LRRTM2. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 26045 | |
| Store at 4°C in the dark. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?