missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRTM3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76529MFV500
This item is not returnable.
View return policy
Description
LRRTM3 Polyclonal antibody specifically detects LRRTM3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| LRRTM3 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| leucine rich repeat transmembrane neuronal 3, leucine-rich repeat transmembrane neuronal protein 3, MGC131810 | |
| Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human LRRTM3. The immunogen is located within the last 50 amino acids of LRRTM3. Amino Acid Squence: SFETNAQEDTMETHLETE | |
| 0.1 mL | |
| Neuroscience | |
| 347731 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction