missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRTM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521101
This item is not returnable.
View return policy
Description
A suggested positive control is Hela cell lysate. PA5-21101 can be used with blocking peptide PEP-1215.
The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Little is known about the function of LRRTM4; in mouse, it is expressed in the limb mesenchyme, neural tube, caudal mesoderm and in three distinct regions of the head. Later expression occurs in a subset of the developing sclerotome. Its similarity to other LRRTM family members suggests that LRRTM4 may also play a role in the development and maintenance of the vertebrate nervous system.
Specifications
| LRRTM4 | |
| Polyclonal | |
| Unconjugated | |
| LRRTM4 | |
| 7530419J18Rik; A230052N11; FLJ12568; leucine rich repeat transmembrane neuronal 4; leucine-rich repeat transmembrane neuronal 4 protein; leucine-rich repeat transmembrane neuronal protein 4; LRRTM4; MGC120633; MGC120636; RGD1560707; UNQ3075/PRO9907 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80059 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q86VH4 | |
| LRRTM4 | |
| A 16 amino acid peptide near the center of human LRRTM4. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction