missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRTM1 Antibody (2381D) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10046JF585
This item is not returnable.
View return policy
Description
LRTM1 Monoclonal antibody specifically detects LRTM1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry,CyTOF
Specifications
| LRTM1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| HT017, leucine-rich repeat and transmembrane domain-containing protein 1, leucine-rich repeats and transmembrane domains 1 | |
| Synthetic peptide containing human LRTM1, Accession # Q9HBL6 | |
| RUO | |
| 57408 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry, CyTOF | |
| 2381D | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction