missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRWD1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97339AF594
This item is not returnable.
View return policy
Description
LRWD1 Polyclonal specifically detects LRWD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| LRWD1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| DKFZp434K1815, leucine-rich repeats and WD repeat domain containing 1, ORCAleucine-rich repeat and WD repeat-containing protein 1, ORC-associated protein, origin recognition complex associated, Origin recognition complex-associated protein | |
| Produced in rabbits immunized with E. coli-derived Human LRWD1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 222229 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction