missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRWD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97339-100ul
This item is not returnable.
View return policy
Description
LRWD1 Polyclonal antibody specifically detects LRWD1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| LRWD1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| DKFZp434K1815, leucine-rich repeats and WD repeat domain containing 1, ORCAleucine-rich repeat and WD repeat-containing protein 1, ORC-associated protein, origin recognition complex associated, Origin recognition complex-associated protein | |
| Produced in rabbits immunized with E. coli-derived Human LRWD1 fragment. | |
| 100 μg | |
| Cell Biology | |
| 222229 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction