missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM1 Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503105
This item is not returnable.
View return policy
Description
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.Specifications
| LSM1 | |
| Monoclonal | |
| 0.35 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15116 | |
| LSM1 | |
| Full length human recombit protein of human LSM1 produced in HEK293 cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1F5 | |
| Unconjugated | |
| LSM1 | |
| CASM, YJL124C | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27257 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction