missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17179
This item is not returnable.
View return policy
Description
LSM11 Polyclonal specifically detects LSM11 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| LSM11 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| LSM11 | |
| Recombinant protein encompassing a sequence within the center region of human LSM11. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 134353 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| 2210404M20Rik, FLJ38273, LSM11 homolog, U7 small nuclear RNA associated, LSM11 homolog, U7 small nuclear RNA associated (S. cerevisiae), LSM11, U7 small nuclear RNA associated, U7 snRNA-associated Sm-like protein LSm11 | |
| Rabbit | |
| 40 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction