missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM11 Monoclonal Antibody (OTI1F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809990
This item is not returnable.
View return policy
Description
LSM11 is a component of the U7 snRNP complex that is involved in the histone 3′-end pre-mRNA processing. Increases U7 snRNA levels but not histone 3′-end pre-mRNA processing activity, when overexpressed. Required for cell cycle progression from G1 to S phases. Binds specifically to the Sm-binding site of U7 snRNA.Specifications
| LSM11 | |
| Monoclonal | |
| Unconjugated | |
| LSM11 | |
| LSM11, U7 small nuclear RNA associated | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 134353 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI1F1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P83369 | |
| LSM11 | |
| Full length human recombit protein of human LSM11 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction