Learn More
LSM11 Monoclonal Antibody (OTI1F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809990S
Encompass_Preferred
 View more versions of this product
Product Code. 17502714
  • € 242.00 / 30µL
Description
LSM11 Monoclonal antibody specifically detects LSM11 in Human samples. It is validated for Western Blot
Specifications
Specifications
LSM11
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P78545
Mouse
Affinity chromatography
RUO
1999
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI1F1
Unconjugated
ELF3
2210404M20Rik; AV079530; FLJ38273; Lsm11; LSM11, U7 small nuclear RNA associated; RGD1565523; U7 snRNA-associated Sm-like protein LSm11; U7 snRNP-specific Sm-like protein LSM11
Human recombinant protein fragment corresponding to amino acids 175-286 of human ELF3 produced in E.coli.