Learn More
LSM11 Monoclonal Antibody (OTI2G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810004S
Encompass_Preferred
 View more versions of this product
Product Code. 17542734
  • € 242.00 / 30µL
Description
LSM11 Monoclonal antibody specifically detects LSM11 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
LSM11
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13287
Mouse
Affinity chromatography
RUO
9111
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2G7
Unconjugated
NMI
2210404M20Rik; AV079530; FLJ38273; Lsm11; LSM11, U7 small nuclear RNA associated; RGD1565523; U7 snRNA-associated Sm-like protein LSm11; U7 snRNP-specific Sm-like protein LSM11
Human recombinant protein fragment corresponding to amino acids 1-223 of human NMI produced in E.coli.