missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12550-100ul
This item is not returnable.
View return policy
Description
LSM12 Polyclonal antibody specifically detects LSM12 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| LSM12 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1-5ul/mg of lysate | |
| FLJ30656, LSM12 homolog (S. cerevisiae), MGC104211, PNAS-135, protein LSM12 homolog | |
| Produced in rabbits immunized with E. coli-derived Human LSM12 fragment | |
| 100 μg | |
| Cell Biology | |
| 124801 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction