missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM3 Monoclonal Antibody (01)

LSM3 Monoclonal Antibody (1)
Brand: Invitrogen MA530653
This item is not returnable.
View return policy
Description
Specifications
| LSM3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P62310 | |
| LSM3 | |
| Recombinant Human LSM3 Protein (Met1-Gly102). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 01 | |
| Unconjugated | |
| LSM3 | |
| 1010001J12Rik; 2610005D18Rik; 6030401D18Rik; Lsm3; LSM3 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM3 homolog, U6 small nuclear RNA associated; LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM3 U6 small nuclear RNA and mRNA degradation associated; MDS017; SMX4; U6 snRNA-associated Sm-like protein LSm3; USS2; YLR438C | |
| Mouse | |
| Protein A | |
| RUO | |
| 27258 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction