missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LSM4 Polyclonal Antibody

Product Code. 15837503
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15837503 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15837503 Supplier Invitrogen™ Supplier No. PA569091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rat: 93%.

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LSM4
Gene Accession No. Q9Y4Z0
Gene Alias Glycine-rich protein; GRP; LSM4; LSM4 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM4 homolog, U6 small nuclear RNA associated; LSM4 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM4 U6 small nuclear RNA and mRNA degradation associated; U6 snRNA-associated SM-like protein 4; U6 snRNA-associated Sm-like protein LSm4; YER112W
Gene Symbols LSM4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human LSM4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25804
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.