Learn More
Invitrogen™ LSM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596467
Description
Immunogen sequence: MLPLSLLKTA QNHPMLVELK NGETYNGHLV SCDNWMNINL REVICTSRDG DKFWRMPECY IRGSTIKYLR IPDEIIDMVK EEVVAKGRGR GGLQQQKQQK GRGMGGAGRG VFGGRGRGGI PGTGRGQPEK KPGRQAGKQ; Positive Samples: HeLa, 22Rv1, HepG2, MCF7, Jurkat, Mouse kidney, Mouse heart, Mouse thymus, Rat liver; Cellular Location: Nucleus.
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
Specifications
| LSM4 | |
| Polyclonal | |
| Unconjugated | |
| LSM4 | |
| Glycine-rich protein; GRP; LSM4; LSM4 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM4 homolog, U6 small nuclear RNA associated; LSM4 homolog, U6 small nuclear RNA associated (S. cerevisiae); LSM4 U6 small nuclear RNA and mRNA degradation associated; U6 snRNA-associated SM-like protein 4; U6 snRNA-associated Sm-like protein LSm4; YER112W | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25804, 290647, 50783 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 2.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9QXA5, Q9Y4Z0 | |
| LSM4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-139 of human LSM4 (NP_0364531). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.