missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LSM6 Monoclonal Antibody (4B5-1B10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524438
This item is not returnable.
View return policy
Description
Peptide Sequence: MSLRKQTPSD FLKQIIGRPV VVKLNSGVDY RGVLACLDGY MNIALEQTEE YVNGQLKNKY GDAFIRGNNV LYISTQKRRM.
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
Specifications
| LSM6 | |
| Monoclonal | |
| 0.49 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P62312 | |
| LSM6 | |
| LSM6 (AAH16026, 1 a.a. ~ 80 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 4B5-1B10 | |
| Unconjugated | |
| LSM6 | |
| LSM6; LSM6 homolog, U6 small nuclear RNA and mRNA degradation associated; LSM6 homolog, U6 small nuclear RNA associated; LSM6 U6 small nuclear RNA and mRNA degradation associated; Sm protein F; U6 snRNA-associated Sm-like protein LSm6; YDR378C | |
| Mouse | |
| Protein A | |
| RUO | |
| 11157 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction