missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesp1 Antibody (939826) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB92193B
This item is not returnable.
View return policy
Description
Mesp1 Monoclonal antibody specifically detects Mesp1 in Human samples. It is validated for Western Blot
Specifications
| Mesp1 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 939826 | |
| Western Blot | |
| BHLHC5, bHLHc5MGC10676, Class C basic helix-loop-helix protein 5, mesoderm posterior 1 homolog (mouse), mesoderm posterior protein 1 | |
| E. coli-derived recombinant human MESP-1, Met1-Gln85, Accession # Q9BRJ9 | |
| 0.1 mL | |
| Cardiovascular Biology, Developmental Biology, Stem Cells | |
| 55897 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction