missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00
Specifications
| Antigen | LSM7 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
LSM7 Polyclonal specifically detects LSM7 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| LSM7 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 51690 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RSGILKGFDPLLNLVLDGTIEYMRDPDDQYKLTEDTRQLGLVVCRGTSVVLICPQDGMEAIPNPFIQQQD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| LSM7 homolog, U6 small nuclear RNA associated (S. cerevisiae), U6 snRNA-associated Sm-like protein LSm7, YNL147W | |
| LSM7 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title