missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LST1 Antibody (LST1/02), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-45072AF647
This item is not returnable.
View return policy
Description
LST1 Monoclonal specifically detects LST1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| LST1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| B144D6S49ELST-1, leucocyte specific transcript 1, leukocyte specific transcript 1, leukocyte-specific transcript 1 protein, lymphocyte antigen 117, MGC119006, MGC119007, Protein B144 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody LST1/02 reacts with an extracellular epitope of LST1, an approximately 6-11 kDa protein expressed as various transmembrane or soluble forms. LST1 is found predominantly on monocytes and dendritic cells. It migrates on SDS PAGE gels as approximately 25-28 kDa molecule. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LST1/02 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| LST1 | |
| KLH-fused peptide corresponding to amino acids SSEGPDLRGRDKRGT of human LST1 | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 7940 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction