missing translation for 'onlineSavingsMsg'
Learn More

LTA4H Mouse anti-Human, Clone: OTI7D11, liquid, TrueMAB™

Product Code. 17582011
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17582011 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17582011 Supplier Origene Technologies Supplier No. TA500643S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.

Specifications

Antigen LTA4H
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone OTI7D11
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LTA4H
Gene Accession No. P09960
Gene Alias leukotriene A4 hydrolase
Gene Symbols LTA4H
Host Species Mouse
Immunogen Full length human recombit protein of human LTA4H produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4048
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.