Learn More
Invitrogen™ LTA4H Recombinant Rabbit Monoclonal Antibody (455)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530654
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Leukotriene A4 Hydrolase/LTA4H.
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
Specifications
| LTA4H | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P09960 | |
| LTA4H | |
| Recombinant Human Leukotriene A4 Hydrolase/LTA4H protein (Met1-Asp611). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 455 | |
| Unconjugated | |
| LTA4H | |
| Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4048 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.