missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTB4R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115525
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LT4R1.
LTB4R (Leukotriene B4 Receptor) is a member of the GPCR family. It is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response. It has been reported in blood, bone marrow, brain, heart, liver, and skeletal muscle. In addition, it has been shown to be highly expressed in leukocytes and lymphoid tissues such as spleen, thymus, and lymph node. ESTs have been isolated from B-cell/lung/testis, breast, colon, eye, kidney, skin, spleen, testis, and tonsil libraries.
Specifications
| LTB4R | |
| Polyclonal | |
| Unconjugated | |
| LTB4R | |
| BLT; BLT1; BLT-1; BLTR; Chemoattractant receptor-like 1; chemokine receptor-like 1; CMKRL1; G protein-coupled receptor 16; GPR16; G-protein coupled receptor 16; leukotriene B4 receptor; leukotriene B4 receptor 1; LTB4 receptor; Ltb4r; LTB4-R 1; Ltb4r1; LTB4-R1; LTBR1; mBLTR; P2RY7; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1241, 16995, 59264 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O88855, Q15722, Q9R0Q2 | |
| LTB4R, Ltb4r1 | |
| A synthesized peptide derived from human LTB4R(Accession Q15722), corresponding to amino acid residues E185-L235. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction