Learn More
Cytiva Luer Connectors for Chromatography Columns
Shop All Cytiva Products
Click to view available options
Product Type:
Tubing Connector
Union, Bottom
Union, Top
Unit Size:
Pack of 2
Description
Requires
Cytiva HiTrap Columns, sold separately.
- HiTrap column inlets have 1/16 in. female threads, while the outlets have 1/16 in. male threads
Specifications
Specifications
| Product Type | Union, Top |
| Quantity | 2 Pack |
| Material | Polypropylene |
| Column Type | Column Accesory |
| For Use With (Application) | Connect a syringe to the top of a HiTrap column |