Learn More
Lufenuron, TRC
Specifications
Specifications
| Chemical Name or Material | Lufenuron |
| CAS | 103055-07-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H8 Cl2 F8 N2 O3 |
| Quantity | 10 g |
| Synonym | Lufenuron anhydrous, 1-[2,5-Dichloro-4-[(2RS)-1,1,2,3,3,3-hexafluoropropoxy]phenyl]-3-(2,6-difluorobenzoyl)urea, Benzamide, N-[[[2,5-dichloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenyl]amino]carbonyl]-2,6-difluoro-, (RS)-1-[2,5-Dichloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenyl]-3-(2,6-difluorobenzoyl)urea, CGA 184699, Cigna, Fluphenacur, Fuoro, Lufenuron, Match, Match (pesticide), Match 5EC, Program |
| InChI Formula | InChI=1S/C17H8Cl2F8N2O3/c18-6-5-11(32-17(26,27)14(22)16(23,24)25)7(19)4-10(6)28-15(31)29-13(30)12-8(20)2-1-3-9(12)21/h1-5,14H,(H2,28,29,30,31) |
| SMILES | FC(C(F)(F)F)C(F)(F)Oc1cc(Cl)c(NC(=O)NC(=O)c2c(F)cccc2F)cc1Cl |
| IUPAC Name | N-[[2,5-dichloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenyl]carbamoyl]-2,6-difluorobenzamide |
| Molecular Weight (g/mol) | 511.15 |
| Show More |