missing translation for 'onlineSavingsMsg'
Learn More

Lufenuron, TRC

Product Code. 30468313
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468313 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468313 Supplier Toronto Research Chemicals Supplier No. TRCL4737501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lufenuron
CAS 103055-07-8
Purity Grade Notes HPLC
Molecular Formula C17 H8 Cl2 F8 N2 O3
Quantity 1 g
Synonym Lufenuron anhydrous, 1-[2,5-Dichloro-4-[(2RS)-1,1,2,3,3,3-hexafluoropropoxy]phenyl]-3-(2,6-difluorobenzoyl)urea, Benzamide, N-[[[2,5-dichloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenyl]amino]carbonyl]-2,6-difluoro-, (RS)-1-[2,5-Dichloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenyl]-3-(2,6-difluorobenzoyl)urea, CGA 184699, Cigna, Fluphenacur, Fuoro, Lufenuron, Match, Match (pesticide), Match 5EC, Program
InChI Formula InChI=1S/C17H8Cl2F8N2O3/c18-6-5-11(32-17(26,27)14(22)16(23,24)25)7(19)4-10(6)28-15(31)29-13(30)12-8(20)2-1-3-9(12)21/h1-5,14H,(H2,28,29,30,31)
SMILES FC(C(F)(F)F)C(F)(F)Oc1cc(Cl)c(NC(=O)NC(=O)c2c(F)cccc2F)cc1Cl
IUPAC Name N-[[2,5-dichloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenyl]carbamoyl]-2,6-difluorobenzamide
Molecular Weight (g/mol) 511.15
Formula Weight 509.9784
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.