Learn More
Lugol Solution, Chem Lab™
Specifications
Specifications
| Color | Brown |
| Boiling Point | 100°C |
| Melting Point | 0°C |
| Composition | 50 g I2 + 100 g KI / l H2O |
| Density | 1.2g/mL |
| Solubility Information | Water Soluble |
| Odor | Iodine |
| pH | ±7 |
| Quantity | 500 mL |
| Physical Form | Liquid |