missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lulizumab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542032
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt. Isotype of this product is: V, kappa.
Lulizumab pegol is under investigation in clinical trial NCT02843659, Proof of Concept Study to Evaluate the Efficacy and Safety of BMS-931699 (Lulizumab) or BMS-986142 in Primary Sjogren's Syndrome.
Specifications
| Lulizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BMS-931699 | |
| Human CD28. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA | |
| 0.51 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| Ig |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction