Learn More
Merck Immobilon Western HRP Chemiluminescence Substrates
Shop All Merck Products
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
Product Type:
HRP Substrate
Sensitivity:
∼1 to 3 pg
∼400 fg
∼6 pg
missing translation for 'unitSize'
500mL
Each
Description
Horseradish peroxidase (HRP) catalyzes the oxidation of luminol in the presence of peroxide, releasing photons (chemiluminescence) that are captured on x-ray films or by CCD cameras. Most HRP substrates are plagued by quickly decaying signal and short shelf life, but Luminata substrates are formulated differently to include stabilizing reagents.
- Consistent results with less pipetting error
The stabilizers added to Luminata substrates:
- Prevent spontaneous oxidation of luminol by peroxide in the absence of HRP
- Enhance the stability of both the luminol and peroxide, ensuring that Luminata substrates are stable for one year in their premixed format
- Stabilize, strengthen, and prolong the chemiluminescence signal once HRP starts to catalyze oxidation
Three different sensitivities for a range of detection needs:
- Immobilon Classico premixed reagent for everyday detection needs
- Immobilon Crescendo premixed reagent for detection in the picogram range
- Immobilon Forte premixed reagent for detection in the femtogram range
Specifications
Specifications
| Product Type | HRP Substrate |
| For Use With (Application) | Dot/Slot Blot |
| Quantity | 500 mL |
| Sensitivity | ∼6 pg |
| Content And Storage | 2°C to 8°C |