missing translation for 'onlineSavingsMsg'
Learn More

Luminespib, MedChemExpress

Product Code. 16416457
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
25mg
500mg
5mg
This item is not returnable. View return policy

Product Code. 16416457

Brand: Medchem Express HY10215

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Luminespib (VER-52296) is a potent HSP90 inhibitor with IC50s of 7.8 and 21 nM for HSP90α and HSP90β, respectively.

Luminespib (VER-52296) is a potent HSP90 inhibitor with IC50s of 7.8 and 21 nM for HSP90α and HSP90β, respectively.

Specifications

Chemical Name or Material Luminespib
CAS 747412-49-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H31N3O5
Quantity 5 mg
Synonym VER-52296 AUY922 NVP-AUY922
Solubility Information DMSO : ≥ 62 mg/mL (133.18 mM)
SMILES O=C(C1=NOC(C2=CC(C(C)C)=C(O)C=C2O)=C1C3=CC=C(CN4CCOCC4)C=C3)NCC
Molecular Weight (g/mol) 465.54
Formula Weight 465.54
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.