missing translation for 'onlineSavingsMsg'
Learn More

Hach™ LUMIStox Luminescent Bacteria Cuvette Tests

Product Code. 10526084
Click to view available options
No. of Tests:
20 Determinations
Includes:
Fully equipped box w/ 20 tubes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10526084

Brand: Hach™ LCK488

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Insert, read and finish. Hach™ LUMIStox Luminescent Bacteria Cuvette Tests are packaged in small tubes of pre-dosed solution in a fully equipped box. They provide maximum safety, ease of use and photometric evaluation for operational analysis.

Hach Cuvette Tests are officially approved for legally required consent limits and provide the assurance that analytical results are always correct. The photometer identifies the cuvette test from its barcode (IBR+) and calculates the result immediately. Cuvettes provide precision and safety using the Dosicap Zip that eliminated contact with the solution.

  • Pre-dosed, ready-to-use solution with no rinsing steps needed
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system
  • Convenient and error-free dosing
  • Barcode label for automatic recognition in the photometer

Specifications

Analytes Luminescent bacteria
Certifications/Compliance EN ISO 11348 part 2
Format Pre-dosed Tubes
For Use With (Application) Water quality testing
For Use With (Equipment) Photometer, including LUMIStox 300
Includes Fully equipped box w/ 20 tubes
No. of Tests 20 Determinations
Test Method Toxicity
Type LUMISTOX Luminescent Bacteria test
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.