missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LUZP4 Antibody (OTI6D11), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72549IR
This item is not returnable.
View return policy
Description
LUZP4 Monoclonal specifically detects LUZP4 in Human samples. It is validated for Western Blot.
Specifications
| LUZP4 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LUZP4 | |
| Full length recombinant protein of human LUZP4 (NP_057467) produced in E.coli. | |
| 0.1 mL | |
| 51213.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI6D11 | |
| Western Blot | |
| leucine zipper protein 4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction