missing translation for 'onlineSavingsMsg'
Learn More

LXH254, MedChemExpress

Product Code. p-200098530
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493126

Brand: Medchem Express HY112089

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LXH254 is a potent, selective, orally active, type II BRAF and CRAF inhibitor, with IC50 values of 0.072 and 0.21 nM against CRAF and BRAF, respectively.

LXH254 is a potent, selective, orally active, type II BRAF and CRAF inhibitor, with IC50 values of 0.072 and 0.21 nM against CRAF and BRAF, respectively.

Specifications

Chemical Name or Material LXH254
CAS 1800398-38-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H25F3N4O4
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (199.01 mM; Need ultrasonic)
SMILES CC1=CC=C(NC(C2=CC(C(F)(F)F)=NC=C2)=O)C=C1C3=CC(N4CCOCC4)=NC(OCCO)=C3
Molecular Weight (g/mol) 502.49
Formula Weight 502.49
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.