missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR alpha/NR1H3 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77106FR
This item is not returnable.
View return policy
Description
LXR alpha/NR1H3 Polyclonal antibody specifically detects LXR alpha/NR1H3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| LXR alpha/NR1H3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus human LXR-A. The immunogen is located within amino acids 50 - 100 of LXR-A. Amino Acid Squence: CTRAEPPSEPTEIRPQ | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators | |
| 10062 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction