missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR alpha/NR1H3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-157JF549
This item is not returnable.
View return policy
Description
LXR alpha/NR1H3 Polyclonal antibody specifically detects LXR alpha/NR1H3 in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Simple Western
Specifications
| LXR alpha/NR1H3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Primate | |
| Purified |
| Western Blot, Simple Western | |
| Janelia Fluor 549 | |
| alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
| A synthetic peptide made to an internal portion of the human LXR alpha/NR1H3 protein sequence (between residues 50-150). [UniProt# Q13133] | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators | |
| 10062 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction