missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR beta/NR1H2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 610.00
Specifications
| Antigen | LXR beta/NR1H2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Form | Purified |
Description
LXR beta/NR1H2 Polyclonal antibody specifically detects LXR beta/NR1H2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| LXR beta/NR1H2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus human LXR-B. The immunogen is located within the first 50 amino acids of LXR-B. Amino Acid Squence: SPTVKEEGPEPWPG | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| mFluor Violet 610 SE | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism | |
| 50mM Sodium Borate | |
| 7376 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title