missing translation for 'onlineSavingsMsg'
Learn More

LY-2584702 tosylate salt, MedChemExpress

Product Code. p-200096587
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16435208

Brand: Medchem Express HY12493A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY-2584702 tosylate salt is a selective ATP competitive inhibitor of p70S6K with an IC50 of 4 nM. In S6K1 enzyme assay, the IC50 of LY-2584702 is 2 nM.

LY-2584702 tosylate salt is a selective ATP competitive inhibitor of p70S6K with an IC50 of 4 nM. In S6K1 enzyme assay, the IC50 of LY-2584702 is 2 nM.

Specifications

Chemical Name or Material LY-2584702 tosylate salt
CAS 1082949-68-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H27F4N7O3S
Quantity 10 mg
Solubility Information DMSO : 10.25 mg/mL (16.60 mM; Need ultrasonic and warming)
SMILES CN1C=C(C2=CC=C(F)C(C(F)(F)F)=C2)N=C1C3CCN(C4=C5C(NN=C5)=NC=N4)CC3.O=S(C6=CC=C(C)C=C6)(O)=O
Molecular Weight (g/mol) 617.62
Formula Weight 617.62
Percent Purity 96.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.