missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-49F Monoclonal Antibody (HBF-719), Biotin

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528665
This item is not returnable.
View return policy
Description
Ly-49F Monoclonal Antibody for Flow
Ly49 receptors (Lectin like receptor) exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
Specifications
| Ly-49F | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.4 | |
| Q60653 | |
| Klra6 | |
| CHO-K1 cells transfected with B6 allele of Ly49F gene. | |
| 1 mL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| HBF-719 | |
| Biotin | |
| Klra6 | |
| Killer cell lectin-like receptor 6; killer cell lectin-like receptor, subfamily A, member 6; Klra6; Ly-49; Ly49F; Ly-49f; Ly49-f; lymphocyte antigen 49f; T-cell surface glycoprotein Ly-49F | |
| Mouse | |
| Purified | |
| RUO | |
| 16637 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction